Product Details

SNP ID
rs61757978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21760769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGGTAAACCTGGTGCAACCCTG[A/G]AGGACTTGCTCTGGATTTCCATTTG
Phenotype
MIM: 176948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK1 PubMed Links

Gene Details

Gene
MAPK1
Gene Name
mitogen-activated protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002745.4 4804 UTR 3 NP_002736.3
NM_138957.3 4804 Intron NP_620407.1

View Full Product Details