Product Details

SNP ID
rs55961300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46245622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGGCCACCTGGTGCTGGGAGC[A/G]GCAGGGGACAGGAAGGACAGCCCCA
Phenotype
MIM: 170998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDPF1 PubMed Links

Gene Details

Gene
CDPF1
Gene Name
cysteine rich DPF motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207327.4 Intron NP_997210.3
XM_011529960.1 Intron XP_011528262.1
XM_011529962.2 Intron XP_011528264.1
XM_011529963.1 Intron XP_011528265.1
XM_011529964.1 Intron XP_011528266.1
XM_011529965.2 Intron XP_011528267.1
Gene
PPARA
Gene Name
peroxisome proliferator activated receptor alpha
There are no transcripts associated with this gene.

View Full Product Details