Product Details
- SNP ID
-
rs61753358
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:55623753 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGACTGTGAATTTTGGTATAGTCT[C/T]CATGGCCAGGTCCCAGGGCTGCTGG
- Phenotype
-
MIM: 300949
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXR2
PubMed Links
Gene Details
- Gene
- FOXR2
- Gene Name
- forkhead box R2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198451.3 |
354 |
Silent Mutation |
CTC,CTT |
L14L |
NP_940853.1 |
View Full Product Details