Product Details

SNP ID
rs62305727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87338466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATTAGGGCAATGATAATAATA[A/G]TAGTAGTAGTTATAATAACAGTGGC
Phenotype
MIM: 612831
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD17B11 PubMed Links

Gene Details

Gene
HSD17B11
Gene Name
hydroxysteroid 17-beta dehydrogenase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016245.4 Intron NP_057329.3
XM_011532021.1 Intron XP_011530323.1

View Full Product Details