Product Details
- SNP ID
-
rs56251804
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:99124464 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCAAAAGATGAGGATTGTTCGG[A/C]CGCTGTTAATAACAATAAAACATTA
- Phenotype
-
MIM: 103740
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADH4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1126673] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADH4
- Gene Name
- alcohol dehydrogenase 4 (class II), pi polypeptide
- Gene
- LOC100507053
- Gene Name
- uncharacterized LOC100507053
There are no transcripts associated with this gene.
View Full Product Details