Product Details
- SNP ID
-
rs56763427
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:100187719 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTGTTCCAATCAGTGAGTAAAG[C/T]TTTTTCTTAACAAGTCGAAATCCTG
- Phenotype
-
MIM: 607730
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DDIT4L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201713115] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDIT4L
- Gene Name
- DNA damage inducible transcript 4 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145244.3 |
785 |
Silent Mutation |
AAA,AAG |
K180K |
NP_660287.1 |
- Gene
- LOC101929353
- Gene Name
- uncharacterized LOC101929353
There are no transcripts associated with this gene.
View Full Product Details