Product Details

SNP ID
rs58796033
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:125317438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCGCGCCGAGGCGCTGATTCAG[C/T]TGCTGGACGTGAATGACAATGACCC
Phenotype
MIM: 612411
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAT4 PubMed Links

Gene Details

Gene
FAT4
Gene Name
FAT atypical cadherin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291285.1 1027 Silent Mutation CTG,TTG L343L NP_001278214.1
NM_001291303.1 1027 Silent Mutation CTG,TTG L343L NP_001278232.1
NM_024582.4 1027 Silent Mutation CTG,TTG L343L NP_078858.4
XM_011532236.2 1027 Silent Mutation CTG,TTG L343L XP_011530538.1
XM_011532237.2 1027 Intron XP_011530539.1

View Full Product Details