Product Details

SNP ID
rs59042568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152916626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAAGGCAGCAGGACCCACTAAA[G/T]CTTTCCTCTTGCACCAGGGTTGCAA
Phenotype
MIM: 605928
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARFIP1 PubMed Links

Gene Details

Gene
ARFIP1
Gene Name
ADP ribosylation factor interacting protein 1
There are no transcripts associated with this gene.

Gene
FHDC1
Gene Name
FH2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033393.2 Intron NP_203751.2
XM_005263319.3 Intron XP_005263376.1
XM_006714393.3 Intron XP_006714456.1
XM_011532388.1 Intron XP_011530690.1
XM_011532389.1 Intron XP_011530691.1
XM_017008787.1 Intron XP_016864276.1

View Full Product Details