Product Details

SNP ID
rs56359437
Assay Type
Functionally Tested
NCBI dbSNP Submissions
29
Location
Chr.4:165331957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCTGTCTTCCGTAACACTCTCC[C/T]CTACCCCGTCGCCACTCTGTCTTCC
Phenotype
MIM: 605774 MIM: 607545
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KLHL2 PubMed Links

Gene Details

Gene
KLHL2
Gene Name
kelch like family member 2
There are no transcripts associated with this gene.

Gene
MSMO1
Gene Name
methylsterol monooxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017369.2 Intron NP_001017369.1
NM_006745.4 Intron NP_006736.1
XM_005263176.2 Intron XP_005263233.1

View Full Product Details