Product Details

SNP ID
rs61942383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106961202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCAAATATGTGGGACTACAGG[C/T]GTGCACCACCGTGCCCAGTCAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM263 PubMed Links

Gene Details

Gene
TMEM263
Gene Name
transmembrane protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319661.1 Intron NP_001306590.1
NM_001319662.1 Intron NP_001306591.1
NM_001319663.1 Intron NP_001306592.1
NM_001319664.1 Intron NP_001306593.1
NM_001319666.1 Intron NP_001306595.1
NM_152261.3 Intron NP_689474.1
XM_017020198.1 Intron XP_016875687.1

View Full Product Details