Product Details

SNP ID
rs59842252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3648199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTTTGTCCTCTTTTCCATTTCT[C/T]CCCCCATTAAGGAGCAGAGGTAATT
Phenotype
MIM: 612396 MIM: 612502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALLC PubMed Links

Gene Details

Gene
ALLC
Gene Name
allantoicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018436.3 Intron NP_060906.3
XM_011510369.2 Intron XP_011508671.1
XM_011510370.2 Intron XP_011508672.1
XM_017004495.1 Intron XP_016859984.1
XM_017004496.1 Intron XP_016859985.1
XM_017004497.1 Intron XP_016859986.1
XM_017004498.1 Intron XP_016859987.1
Gene
COLEC11
Gene Name
collectin subfamily member 11
There are no transcripts associated with this gene.

View Full Product Details