Product Details

SNP ID
rs62166769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85585661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTGAAGGCTGAAGGCAGGATA[A/T]TTTATTGGGGGAGGAGGATACTGGG
Phenotype
MIM: 612490 MIM: 607029 MIM: 603177
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF181 PubMed Links

Gene Details

Gene
RNF181
Gene Name
ring finger protein 181
There are no transcripts associated with this gene.

Gene
VAMP5
Gene Name
vesicle associated membrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006634.2 Intron NP_006625.1
Gene
VAMP8
Gene Name
vesicle associated membrane protein 8
There are no transcripts associated with this gene.

View Full Product Details