Product Details

SNP ID
rs56226634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51418199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGTCAGCAGCAAGGACCCATC[C/T]TAAAAGAAAAAGGCGCAAGGTGGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF1 PubMed Links

Gene Details

Gene
DCAF1
Gene Name
DDB1 and CUL4 associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171904.1 Intron NP_001165375.1
NM_014703.2 Intron NP_055518.1
XM_005276753.4 Intron XP_005276810.1
XM_005276755.4 Intron XP_005276812.1
XM_011534273.2 Intron XP_011532575.1
XM_011534274.2 Intron XP_011532576.1
XM_011534275.2 Intron XP_011532577.1
XM_011534276.2 Intron XP_011532578.1
XM_011534277.2 Intron XP_011532579.1
XM_017007546.1 Intron XP_016863035.1
XM_017007547.1 Intron XP_016863036.1
XM_017007548.1 Intron XP_016863037.1
XM_017007549.1 Intron XP_016863038.1
XM_017007550.1 Intron XP_016863039.1
XM_017007551.1 Intron XP_016863040.1

View Full Product Details