Product Details

SNP ID
rs61379910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169407238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCATCTTCCTATGGTCAACTTGC[C/T]CCACACAGTCTTGCCCAAATGCTGC
Phenotype
MIM: 604588
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEK1 PubMed Links

Gene Details

Gene
NEK1
Gene Name
NIMA related kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199397.1 Intron NP_001186326.1
NM_001199398.1 Intron NP_001186327.1
NM_001199399.1 Intron NP_001186328.1
NM_001199400.1 Intron NP_001186329.1
NM_012224.2 Intron NP_036356.1
XM_006714228.1 Intron XP_006714291.1
XM_011532003.1 Intron XP_011530305.1
XM_011532004.1 Intron XP_011530306.1
XM_011532005.2 Intron XP_011530307.1
XM_017008249.1 Intron XP_016863738.1
XM_017008250.1 Intron XP_016863739.1
XM_017008251.1 Intron XP_016863740.1
XM_017008252.1 Intron XP_016863741.1
XM_017008253.1 Intron XP_016863742.1
XM_017008254.1 Intron XP_016863743.1

View Full Product Details