Product Details
- SNP ID
-
rs62390520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:181193970 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTTTCATCTCCTCACCCCCTTCC[C/G]AGGGGAGCGCTGAGCAGGGAGTAGC
- Phenotype
-
MIM: 609315
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC102577426
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75186999] are located under a probe and SNP(s) [rs149724261] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC102577426
- Gene Name
- uncharacterized LOC102577426
There are no transcripts associated with this gene.
- Gene
- TRIM7
- Gene Name
- tripartite motif containing 7
View Full Product Details