Product Details

SNP ID
rs61758103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:181224383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTATGTGTGGGAGGAGGAGGA[C/T]GAGGAGGAAGACCTGGACTACTACT
Phenotype
MIM: 610530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4638 PubMed Links

Gene Details

Gene
MIR4638
Gene Name
microRNA 4638
There are no transcripts associated with this gene.

Gene
TRIM41
Gene Name
tripartite motif containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033549.4 1119 Silent Mutation GAC,GAT D128D NP_291027.3
NM_201627.2 1119 Silent Mutation GAC,GAT D128D NP_963921.1
XM_006714929.2 1119 Missense Mutation GAC,GAT D128D XP_006714992.1
XM_006714930.2 1119 Silent Mutation GAC,GAT D128D XP_006714993.1
XM_006714931.3 1119 Missense Mutation GAC,GAT D128D XP_006714994.1
XM_006714932.3 1119 Missense Mutation GAC,GAT D128D XP_006714995.1

View Full Product Details