Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001303618.1 | 2008 | UTR 3 | NP_001290547.1 | ||
NM_001303619.1 | 2008 | UTR 3 | NP_001290548.1 | ||
NM_006566.3 | 2008 | UTR 3 | NP_006557.2 | ||
XM_005266642.3 | 2008 | UTR 3 | XP_005266699.1 | ||
XM_005266643.3 | 2008 | UTR 3 | XP_005266700.1 | ||
XM_006722374.3 | 2008 | UTR 3 | XP_006722437.1 | ||
XM_017025524.1 | 2008 | UTR 3 | XP_016881013.1 | ||
XM_017025525.1 | 2008 | UTR 3 | XP_016881014.1 | ||
XM_017025526.1 | 2008 | UTR 3 | XP_016881015.1 | ||
XM_017025527.1 | 2008 | UTR 3 | XP_016881016.1 |