Product Details
- SNP ID
-
rs62094319
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:58044665 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGCGCGGGGCGCCGGCTCCATGG[A/C]GACCGGGCTCGGGGAGCCGGTCTAT
- Phenotype
-
MIM: 606384
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NEDD4L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113358301] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NEDD4L
- Gene Name
- neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
View Full Product Details