Product Details

SNP ID
rs58797899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59268087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTGGCACCGAACCGGAGATCT[A/G]CTTGGTGAACTGAGAGGAGTCCTTA
Phenotype
MIM: 601881
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAX PubMed Links

Gene Details

Gene
RAX
Gene Name
retina and anterior neural fold homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013435.2 2145 UTR 3 NP_038463.2

View Full Product Details