Product Details

SNP ID
hCV90269446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:67345782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTACTTACCTGCAGAGAAGCTA[A/G]TTCCACCAGTAGTTCCACTGCTTTG
Phenotype
MIM: 609398 MIM: 603930 MIM: 606958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1D PubMed Links

Gene Details

Gene
ATP6V1D
Gene Name
ATPase H+ transporting V1 subunit D
There are no transcripts associated with this gene.

Gene
GPHN
Gene Name
gephyrin
There are no transcripts associated with this gene.

Gene
LOC105370541
Gene Name
uncharacterized LOC105370541
There are no transcripts associated with this gene.

Gene
MPP5
Gene Name
membrane palmitoylated protein 5
There are no transcripts associated with this gene.

View Full Product Details