Product Details

SNP ID
rs60640368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48767154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCCAAAGCATACCATGTCCCA[A/G]GACACTATCTGGGGAAGCCAGGAAA
Phenotype
MIM: 600294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY6 PubMed Links

Gene Details

Gene
ADCY6
Gene Name
adenylate cyclase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015270.4 5301 UTR 3 NP_056085.1
XM_006719210.3 5301 Intron XP_006719273.1
XM_017018743.1 5301 UTR 3 XP_016874232.1
Gene
LINC00935
Gene Name
long intergenic non-protein coding RNA 935
There are no transcripts associated with this gene.

Gene
MIR4701
Gene Name
microRNA 4701
There are no transcripts associated with this gene.

View Full Product Details