Product Details
- SNP ID
-
rs59827092
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49297468 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGGAGGAGCTGCGACAGCTAAAA[A/G]AGGAGATGGCGCGGCACCTGAGGGA
- Phenotype
-
MIM: 170710
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927267
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73112143] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927267
- Gene Name
- uncharacterized LOC101927267
There are no transcripts associated with this gene.
- Gene
- PRPH
- Gene Name
- peripherin
View Full Product Details