Product Details

SNP ID
rs56360558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51954867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCAGTGAATAAGCAATGCTGG[A/G]TTACTTTTTTATCTTAAGGAGCAGA
Phenotype
MIM: 601300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACVR1B PubMed Links

Gene Details

Gene
ACVR1B
Gene Name
activin A receptor type 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004302.4 Intron NP_004293.1
NM_020327.3 Intron NP_064732.3
NM_020328.3 Intron NP_064733.3
XM_011538966.2 Intron XP_011537268.1
XM_011538967.2 Intron XP_011537269.1
XM_017020199.1 Intron XP_016875688.1
XM_017020200.1 Intron XP_016875689.1
XM_017020201.1 Intron XP_016875690.1

View Full Product Details