Product Details

SNP ID
rs56305333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57001977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATATCAAAGTGGTTCCCTATCC[A/T]CTCTGCCCCACAGAAGGCCGGACGT
Phenotype
MIM: 605307 MIM: 162330
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR182 PubMed Links

Gene Details

Gene
GPR182
Gene Name
G protein-coupled receptor 182
There are no transcripts associated with this gene.

Gene
HBCBP
Gene Name
HBcAg-binding protein
There are no transcripts associated with this gene.

Gene
TAC3
Gene Name
tachykinin 3
There are no transcripts associated with this gene.

Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014830.2 3094 UTR 3 NP_055645.1

View Full Product Details