Product Details

SNP ID
rs61771494
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:68429603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAATACGTACTTTTTTTTTTAA[A/T]TAAAGGAATTGCTTGCTCAACTCAG
Phenotype
MIM: 180069
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RPE65 PubMed Links

Gene Details

Gene
RPE65
Gene Name
RPE65, retinoid isomerohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000329.2 2041 UTR 3 NP_000320.1
XM_017002027.1 2041 UTR 3 XP_016857516.1

View Full Product Details