Product Details
- SNP ID
-
rs61905116
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:116778822 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGATGTCCTCCCCACCATGGGCA[A/G]GGGCTGGTGGGAAAGACACTCCCAG
- Phenotype
-
MIM: 603901
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BUD13
PubMed Links
Gene Details
- Gene
- BUD13
- Gene Name
- BUD13 homolog
There are no transcripts associated with this gene.
- Gene
- ZPR1
- Gene Name
- ZPR1 zinc finger
View Full Product Details