Product Details
- SNP ID
-
rs57300701
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118974836 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAAGACAGACAATTGAGTATGAG[C/G]CTTCAAAGTGTTTTGACCTAGTGGA
- Phenotype
-
MIM: 615755
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOXR1
PubMed Links
Gene Details
- Gene
- FOXR1
- Gene Name
- forkhead box R1
View Full Product Details