Product Details

SNP ID
rs55662663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65525992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATATACCTCAAGGCGAGAGTGGA[A/G]GCTGGTGGCCTGAAGGAGCTGGAGA
Phenotype
MIM: 607982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCYL1 PubMed Links

Gene Details

Gene
SCYL1
Gene Name
SCY1 like pseudokinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048218.1 401 Silent Mutation GAA,GAG E108E NP_001041683.1
NM_020680.3 401 Silent Mutation GAA,GAG E108E NP_065731.3
XM_005274118.3 401 Silent Mutation GAA,GAG E108E XP_005274175.1
XM_005274120.3 401 Silent Mutation GAA,GAG E108E XP_005274177.1
XM_005274121.3 401 Silent Mutation GAA,GAG E108E XP_005274178.1
XM_017018029.1 401 Silent Mutation GAA,GAG E108E XP_016873518.1
XM_017018030.1 401 Silent Mutation GAA,GAG E108E XP_016873519.1
XM_017018031.1 401 Silent Mutation GAA,GAG E108E XP_016873520.1
XM_017018032.1 401 UTR 5 XP_016873521.1
XM_017018033.1 401 UTR 5 XP_016873522.1

View Full Product Details