Product Details
- SNP ID
-
rs56102510
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:67628153 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGTGGTCCATGCAGGAAGACGGG[C/T]AGTGTGTAGCGGAAGTGGCCACCCC
- Phenotype
-
MIM: 604648
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUDT8
PubMed Links
Gene Details
- Gene
- NUDT8
- Gene Name
- nudix hydrolase 8
- Gene
- TBX10
- Gene Name
- T-box 10
There are no transcripts associated with this gene.
View Full Product Details