Product Details
- SNP ID
-
rs58520141
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:77069856 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCCTGGGCTCACCGTGGTCCTGA[A/G]GCTTGGCCTTCCTGCTGCCCACCTC
- Phenotype
-
MIM: 602537
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN5
PubMed Links
Gene Details
- Gene
- CAPN5
- Gene Name
- calpain 5
View Full Product Details