Product Details

SNP ID
rs57055606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44294893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGCGTCATAATACCTCCGCCT[C/T]AGTACACTCCCCATCCTGAGGGGCG
Phenotype
MIM: 600835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf142 PubMed Links

Gene Details

Gene
C10orf142
Gene Name
chromosome 10 open reading frame 142
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258000.1 Intron NP_001244929.1
Gene
CXCL12
Gene Name
C-X-C motif chemokine ligand 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000609.6 Intron NP_000600.1
NM_001033886.2 Intron NP_001029058.1
NM_001178134.1 Intron NP_001171605.1
NM_001277990.1 Intron NP_001264919.1
NM_199168.3 Intron NP_954637.1

View Full Product Details