Product Details

SNP ID
rs56002406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44963809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATATGCTTAGAACATTTTAATTT[A/G]GATGCCTGTACGTTTTCCCTTTTGC
Phenotype
MIM: 610559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASSF4 PubMed Links

Gene Details

Gene
RASSF4
Gene Name
Ras association domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032023.3 Intron NP_114412.2
XM_005271833.4 Intron XP_005271890.1
XM_005271835.4 Intron XP_005271892.1
XM_006718013.3 Intron XP_006718076.1
XM_006718014.3 Intron XP_006718077.1
XM_011540255.2 Intron XP_011538557.1
XM_017016754.1 Intron XP_016872243.1
XM_017016755.1 Intron XP_016872244.1
XM_017016756.1 Intron XP_016872245.1
XM_017016757.1 Intron XP_016872246.1
XM_017016758.1 Intron XP_016872247.1
Gene
TMEM72-AS1
Gene Name
TMEM72 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details