Product Details

SNP ID
rs56023848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:113026232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAACTACCATTCCCTCACTCTG[C/T]CCCCGTCATGACAACAGGAGCTAAG
Phenotype
MIM: 609844 MIM: 159350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCP2 PubMed Links

Gene Details

Gene
DCP2
Gene Name
decapping mRNA 2
There are no transcripts associated with this gene.

Gene
MCC
Gene Name
mutated in colorectal cancers
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085377.1 4190 UTR 3 NP_001078846.1
NM_002387.2 4190 UTR 3 NP_002378.1
XM_005271991.2 4190 UTR 3 XP_005272048.1
XM_017009473.1 4190 UTR 3 XP_016864962.1
XM_017009474.1 4190 UTR 3 XP_016864963.1

View Full Product Details