Product Details
- SNP ID
-
rs55848952
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:1394631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCGCCTGCTCACACATGTCGATG[C/T]GGAGTGCCCGCCTGCTCACACATGC
- Phenotype
-
MIM: 610203
MIM: 614632
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRIPAK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78309237] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRIPAK
- Gene Name
- cysteine rich PAK1 inhibitor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175918.3 |
3080 |
Silent Mutation |
TGC,TGT |
C40C |
NP_787114.2 |
- Gene
- NKX1-1
- Gene Name
- NK1 homeobox 1
There are no transcripts associated with this gene.
- Gene
- UVSSA
- Gene Name
- UV stimulated scaffold protein A
There are no transcripts associated with this gene.
View Full Product Details