Product Details

SNP ID
rs56397731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3962824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGTGATTTTCCAAACAACCAA[C/T]TAAAAAAAACTCCTTACCTCGAAAA
Phenotype
MIM: 603302
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCY9 PubMed Links

Gene Details

Gene
ADCY9
Gene Name
adenylate cyclase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001116.3 7434 UTR 3 NP_001107.2
XM_005255079.3 7434 UTR 3 XP_005255136.1
XM_011522353.2 7434 Intron XP_011520655.1

View Full Product Details