Product Details

SNP ID
rs71514277
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:26707937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATACTAACTTTGCTTCTCCAAA[A/G]TAAATTACTCAGGCACCAAGATTGT
Phenotype
MIM: 603024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID1A PubMed Links

Gene Details

Gene
ARID1A
Gene Name
AT-rich interaction domain 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006015.4 Intron NP_006006.3
NM_139135.2 Intron NP_624361.1

View Full Product Details