Product Details

SNP ID
rs72474554
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:15466144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGGCGTGATCTCCAGCTGCAA[C/T]GTGAGTACCAAAATCAGGGGCTCCG
Phenotype
MIM: 609443 MIM: 609444
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELA2A PubMed Links

Gene Details

Gene
CELA2A
Gene Name
chymotrypsin like elastase family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033440.2 665 Silent Mutation AAC,AAT N213N NP_254275.1
Gene
CELA2B
Gene Name
chymotrypsin like elastase family member 2B
There are no transcripts associated with this gene.

View Full Product Details