Product Details

SNP ID
rs73991610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201488716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGATTTGAACACAGGTGGGGAT[A/G]GATTAATAGAACTGGGGATCAGGGA
Phenotype
MIM: 607333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALS2CR11 PubMed Links

Gene Details

Gene
ALS2CR11
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168216.1 Intron NP_001161688.1
NM_001168217.1 Intron NP_001161689.1
NM_001168221.1 Intron NP_001161693.1
NM_152525.5 Intron NP_689738.3
XM_006712331.3 Intron XP_006712394.1
XM_006712332.3 Intron XP_006712395.1
XM_006712333.3 Intron XP_006712396.1
XM_006712334.3 Intron XP_006712397.1
XM_006712335.3 Intron XP_006712398.1
XM_006712336.3 Intron XP_006712399.1
XM_011510736.2 Intron XP_011509038.1
Gene
STRADB
Gene Name
STE20-related kinase adaptor beta
There are no transcripts associated with this gene.

View Full Product Details