Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001270942.1 | 7077 | UTR 3 | NP_001257871.1 | ||
NM_001270943.1 | 7077 | UTR 3 | NP_001257872.1 | ||
NM_001270944.1 | 7077 | UTR 3 | NP_001257873.1 | ||
NM_003709.3 | 7077 | UTR 3 | NP_003700.1 | ||
XM_005246926.3 | 7077 | Intron | XP_005246983.1 | ||
XM_006712815.3 | 7077 | Intron | XP_006712878.1 | ||
XM_011512072.2 | 7077 | Intron | XP_011510374.1 | ||
XM_011512073.2 | 7077 | Intron | XP_011510375.1 | ||
XM_011512074.2 | 7077 | Intron | XP_011510376.1 | ||
XM_011512075.1 | 7077 | Intron | XP_011510377.1 | ||
XM_017005161.1 | 7077 | Intron | XP_016860650.1 | ||
XM_017005162.1 | 7077 | Intron | XP_016860651.1 | ||
XM_017005163.1 | 7077 | Intron | XP_016860652.1 | ||
XM_017005164.1 | 7077 | Intron | XP_016860653.1 |