Product Details

SNP ID
rs73056953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207593018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAATATGTTAGGATGTTGGAGA[C/T]GGTCACACAGCTTTTTGTTGTTCTG
Phenotype
MIM: 123810 MIM: 615257
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREB1 PubMed Links

Gene Details

Gene
CREB1
Gene Name
cAMP responsive element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320793.1 Intron NP_001307722.1
NM_004379.4 Intron NP_004370.1
NM_134442.4 Intron NP_604391.1
XM_011510645.1 Intron XP_011508947.1
XM_011510646.2 Intron XP_011508948.1
XM_011510647.2 Intron XP_011508949.1
XM_011510648.2 Intron XP_011508950.1
XM_011510649.2 Intron XP_011508951.1
XM_011510650.2 Intron XP_011508952.1
XM_011510651.2 Intron XP_011508953.1
XM_017003399.1 Intron XP_016858888.1
XM_017003400.1 Intron XP_016858889.1
XM_017003401.1 Intron XP_016858890.1
Gene
METTL21A
Gene Name
methyltransferase like 21A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005246336.3 Intron XP_005246393.1
XM_005246337.4 Intron XP_005246394.1
XM_005246338.4 Intron XP_005246395.1
XM_005246339.3 Intron XP_005246396.1
XM_005246340.3 Intron XP_005246397.1
XM_005246341.2 Intron XP_005246398.1
XM_006712327.3 Intron XP_006712390.1
XM_006712329.3 Intron XP_006712392.1
XM_011510724.2 Intron XP_011509026.1
XM_011510725.2 Intron XP_011509027.1
XM_011510727.2 Intron XP_011509029.1
XM_011510728.2 Intron XP_011509030.1
XM_011510729.2 Intron XP_011509031.1
XM_017003442.1 Intron XP_016858931.1
XM_017003443.1 Intron XP_016858932.1
XM_017003444.1 Intron XP_016858933.1
XM_017003445.1 Intron XP_016858934.1
XM_017003446.1 Intron XP_016858935.1
XM_017003447.1 Intron XP_016858936.1
XM_017003448.1 Intron XP_016858937.1
XM_017003449.1 Intron XP_016858938.1
XM_017003450.1 Intron XP_016858939.1

View Full Product Details