Product Details

SNP ID
rs72711321
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:220414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATTGATTTCTTTTCTTTGAATG[G/T]ATTGTTCTGAATATGTTTAATGGTG
Phenotype
MIM: 600857
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CCDC127 PubMed Links
Additional Information
For this assay, SNP(s) [rs56035544] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CCDC127
Gene Name
coiled-coil domain containing 127
There are no transcripts associated with this gene.

Gene
SDHA
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294332.1 Intron NP_001281261.1
NM_004168.3 Intron NP_004159.2
XM_005248331.3 Intron XP_005248388.1
XM_011514072.1 Intron XP_011512374.1
XM_011514073.1 Intron XP_011512375.1
XM_017009685.1 Intron XP_016865174.1

View Full Product Details