Product Details

SNP ID
rs72758673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43536304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAACCTACAAAAGTATCTGCACA[C/T]ATATATGACAAGAGGAAAGATTACA
Phenotype
MIM: 605184
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAIP1 PubMed Links

Gene Details

Gene
PAIP1
Gene Name
poly(A) binding protein interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006451.4 Intron NP_006442.2
NM_182789.3 Intron NP_877590.1
NM_183323.2 Intron NP_899152.1
XM_005248230.3 Intron XP_005248287.1
XM_017008956.1 Intron XP_016864445.1

View Full Product Details