Product Details

SNP ID
rs73437410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52795947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCATCCGTGTTTGGAAATTACC[A/G]TGAGGTGCTGGATCCATGGACTGCA
Phenotype
MIM: 138359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTA1 PubMed Links

Gene Details

Gene
GSTA1
Gene Name
glutathione S-transferase alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319059.1 Intron NP_001305988.1
NM_145740.4 Intron NP_665683.1
XM_005249034.3 Intron XP_005249091.1

View Full Product Details