Product Details

SNP ID
rs67622831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53649409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCTGAGTCTTCAGATAAACAA[C/T]GTAGAAAATAAATTAAATGGTAATT
Phenotype
MIM: 610749
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL31 PubMed Links

Gene Details

Gene
KLHL31
Gene Name
kelch like family member 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003760.4 4235 UTR 3 NP_001003760.2
XM_005249107.4 4235 Intron XP_005249164.1
XM_017010858.1 4235 Intron XP_016866347.1

View Full Product Details