Product Details

SNP ID
rs73053492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:12690639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGAAGGCTGTTTTTTTTTTTTT[C/T]TCCCTTCACTGATACTGAAATCATG
Phenotype
MIM: 604786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL4A PubMed Links

Gene Details

Gene
ARL4A
Gene Name
ADP ribosylation factor like GTPase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037164.2 2914 UTR 3 NP_001032241.1
NM_001195396.1 2914 UTR 3 NP_001182325.1
NM_005738.4 2914 UTR 3 NP_005729.1
NM_212460.3 2914 UTR 3 NP_997625.1

View Full Product Details