Product Details

SNP ID
rs73063942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:16650732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATAAACAAGAATACTTTTCATT[C/T]TTAACTCACTCATTTCTGTGTATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKMY2 PubMed Links

Gene Details

Gene
ANKMY2
Gene Name
ankyrin repeat and MYND domain containing 2
There are no transcripts associated with this gene.

Gene
BZW2
Gene Name
basic leucine zipper and W2 domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159767.1 Intron NP_001153239.1
NM_014038.2 Intron NP_054757.1
XM_006715706.1 Intron XP_006715769.1
XM_006715707.1 Intron XP_006715770.1
XM_006715708.1 Intron XP_006715771.1

View Full Product Details