Product Details

SNP ID
rs73472731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140334428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGCATCTGTACTTGACACTTA[C/G]TAGAAGAAAAGGTCTTTCCCTTTTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC37A3 PubMed Links

Gene Details

Gene
SLC37A3
Gene Name
solute carrier family 37 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287498.1 2970 UTR 3 NP_001274427.1
NM_032295.3 2970 UTR 3 NP_115671.1
NM_207113.2 2970 UTR 3 NP_996996.1
XM_011516626.2 2970 UTR 3 XP_011514928.1
XM_011516627.2 2970 UTR 3 XP_011514929.1
XM_017012712.1 2970 UTR 3 XP_016868201.1
XM_017012713.1 2970 UTR 3 XP_016868202.1
XM_017012714.1 2970 UTR 3 XP_016868203.1
XM_017012715.1 2970 UTR 3 XP_016868204.1
XM_017012716.1 2970 UTR 3 XP_016868205.1
XM_017012717.1 2970 UTR 3 XP_016868206.1
XM_017012718.1 2970 Intron XP_016868207.1

View Full Product Details