Product Details

SNP ID
rs73445821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135929178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTATATAAATGTATGAATCTA[C/G]AGGAGAAAAAAATATGCATACTGGA
Phenotype
MIM: 611050 MIM: 606484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LUZP6 PubMed Links

Gene Details

Gene
LUZP6
Gene Name
leucine zipper protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128619.2 1461 UTR 5 NP_001122091.2
Gene
MTPN
Gene Name
myotrophin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145808.3 1461 UTR 3 NP_665807.1

View Full Product Details