Product Details

SNP ID
rs73168924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138834938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGAATGAAGTTCAGAGAAGGAA[C/T]CAGACATGGTCAGTGAGTGACAAAG
Phenotype
MIM: 613344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1549 PubMed Links

Gene Details

Gene
KIAA1549
Gene Name
KIAA1549
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164665.1 9097 UTR 3 NP_001158137.1
NM_020910.2 9097 UTR 3 NP_065961.2
XM_011516442.2 9097 UTR 3 XP_011514744.1

View Full Product Details