Product Details

SNP ID
rs72668936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:108444175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAACAAATGTGATTCTCATATGC[C/T]ACTCTACATTTAGTTTTCTTCTTAA
Phenotype
MIM: 607722
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC2 PubMed Links

Gene Details

Gene
EMC2
Gene Name
ER membrane protein complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details